ENA408615468 Enamine

2-[(2-aminoethyl)amino]-N-[(pyridin-2-yl)methyl]acetamide trihydrochloride

  • MDL number MFCD26407775
  • Cas Number : 1461708-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136887
Availability details for
Quantity
  • Availability
  • Add to Cart