ENA408615469 Enamine

1-(3-fluorophenyl)piperazin-2-one hydrochloride

  • MDL number MFCD26407777
  • Cas Number : 914654-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136897

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136897
Availability details for
Quantity
  • Availability
  • Add to Cart