ENA408615471 Enamine

3-(cyclopentyloxy)-N'-hydroxy-4-methoxybenzene-1-carboximidamide hydrochloride

  • MDL number MFCD26407779
  • Cas Number : 1461726-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136904

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136904
Availability details for
Quantity
  • Availability
  • Add to Cart