ENA408615473 Enamine

methyl 2-amino-3-(1,2,3,4-tetrahydronaphthalen-1-yl)propanoate hydrochloride

  • MDL number MFCD26407781
  • Cas Number : 1461705-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136922
Availability details for
Quantity
  • Availability
  • Add to Cart