ENA408615477 Enamine

methyl 4-(benzyloxy)-3-methoxy-2-nitrobenzoate

  • MDL number MFCD26407791
  • Cas Number : 1461706-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137035

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137035
Availability details for
Quantity
  • Availability
  • Add to Cart