ENA408615478 Enamine

(3S)-octahydropyrido[2,1-c][1,4]oxazine-3-carbonitrile

  • MDL number MFCD26407792
  • Cas Number : 1461698-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137036
Availability details for
Quantity
  • Availability
  • Add to Cart