ENA408615479 Enamine

(3R)-octahydropyrido[2,1-c]morpholine-3-carbonitrile

  • MDL number MFCD26407793
  • Cas Number : 1461698-13-4
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3R)-octahydropyrido[2,1-c][1,4]oxazine-3-carbonitrile
Inchi Code 1S/C9H14N2O/c10-5-9-6-11-4-2-1-3-8(11)7-12-9/h8-9H,1-4,6-7H2/t8?,9-/m0/s1
Inchi Key IUYZKCJVSORTEH-GKAPJAKFSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137037
Availability details for
Quantity
  • Availability
  • Add to Cart