ENA408615490 Enamine

N-(prop-2-yn-1-yl)cyclopropanamine hydrochloride

  • MDL number MFCD26407813
  • Cas Number : 64892-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137177

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137177
Availability details for
Quantity
  • Availability
  • Add to Cart