ENA408615496 Enamine

5-bromo-1,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD26407819
  • Cas Number : 450841-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137188
Availability details for
Quantity
  • Availability
  • Add to Cart