ENA408615498 Enamine

1-chloropropan-2-amine hydrochloride

  • MDL number MFCD26407822
  • Cas Number : 5968-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137231

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137231
Availability details for
Quantity
  • Availability
  • Add to Cart