ENA408615499 Enamine

N-{5-[1-(ethoxyamino)-2-nitroethyl]pyridin-2-yl}acetamide

  • MDL number MFCD26407823
  • Cas Number : 1461705-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137234
Availability details for
Quantity
  • Availability
  • Add to Cart