ENA408615509 Enamine

propan-2-yl 4-aminobutanoate hydrochloride

  • MDL number MFCD00465277
  • Cas Number : 64834-27-1
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137382
Availability details for
Quantity
  • Availability
  • Add to Cart