ENA408615524 Enamine

5-chloro-4-acetamido-2-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD26407874
  • Cas Number : 1461705-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137658
Availability details for
Quantity
  • Availability
  • Add to Cart