ENA408615544 Enamine

N-aminobenzenecarbothioamide

  • MDL number MFCD00463358
  • Cas Number : 20605-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134656
Availability details for
Quantity
  • Availability
  • Add to Cart