ENA408615551 Enamine

3-(4-acetylphenyl)-1,2,3$l^{5}-oxadiazol-3-ylium-5-olate

  • MDL number MFCD26407628
  • Cas Number : 84968-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134905

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134905
Availability details for
Quantity
  • Availability
  • Add to Cart