ENA408615553 Enamine

tert-butyl 3-[4-(methoxycarbonyl)-1H-1,2,3-triazol-1-yl]piperidine-1-carboxylate

  • MDL number MFCD26141883
  • Cas Number : 1461715-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134942
Availability details for
Quantity
  • Availability
  • Add to Cart