ENA408615554 Enamine

tert-butyl 3-(methanesulfonyloxy)piperidine-1-carboxylate

  • MDL number MFCD07787193
  • Cas Number : 129888-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134943

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134943
Availability details for
Quantity
  • Availability
  • Add to Cart