ENA408615556 Enamine

2-(propan-2-yl)-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD00183540
  • Cas Number : 41816-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135075
Availability details for
Quantity
  • Availability
  • Add to Cart