ENA408615557 Enamine

6-tert-butyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD10478931
  • Cas Number : 4138-19-6
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-135108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135108
Availability details for
Quantity
  • Availability
  • Add to Cart