ENA408615558 Enamine

2-amino-3-hydroxy-N-methylpropanamide

  • MDL number MFCD19206881
  • Cas Number : 57645-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-135130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135130
Availability details for
Quantity
  • Availability
  • Add to Cart