ENA408615568 Enamine

1-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine hydrochloride

  • MDL number MFCD26407640
  • Cas Number : 1461713-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135331
Availability details for
Quantity
  • Availability
  • Add to Cart