ENA408615572 Enamine

2-(dimethylamino)propan-1-ol

  • MDL number MFCD00014425
  • Cas Number : 15521-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135379

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-135379
pictograms GHS02,GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart