ENA408615580 Enamine

2-[benzyl(2-{[(tert-butoxy)carbonyl]amino}ethyl)amino]acetic acid

  • MDL number MFCD26407642
  • Cas Number : 1461708-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135499

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135499
Availability details for
Quantity
  • Availability
  • Add to Cart