ENA408615589 Enamine

3,3-diethylpyrrolidin-2-one

  • MDL number MFCD07784501
  • Cas Number : 175698-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135573

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135573
Availability details for
Quantity
  • Availability
  • Add to Cart