ENA408615595 Enamine

tert-butyl 2-hydroxybenzoate

  • MDL number MFCD19600053
  • Cas Number : 23408-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135621

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135621
Availability details for
Quantity
  • Availability
  • Add to Cart