ENA408615601 Enamine

pyrido[2,3-b]pyrazine-7-carboxylic acid

  • MDL number MFCD06496468
  • Cas Number : 893723-49-4
  • Molecular Weight : 175.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name pyrido[2,3-b]pyrazine-7-carboxylic acid
Inchi Code 1S/C8H5N3O2/c12-8(13)5-3-6-7(11-4-5)10-2-1-9-6/h1-4H,(H,12,13)
Inchi Key SMPGSEFDPLFZJS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-135662

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-135662
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart