ENA408615604 Enamine

tert-butyl 2-aminohexanoate

  • MDL number MFCD12804802
  • Cas Number : 17389-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-135669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135669
Availability details for
Quantity
  • Availability
  • Add to Cart