ENA408615607 Enamine

tert-butyl 4-[(6-ethoxypyrimidin-4-yl)amino]piperidine-1-carboxylate

  • MDL number MFCD21098547
  • Cas Number : 1353955-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135687
Availability details for
Quantity
  • Availability
  • Add to Cart