ENA408615609 Enamine

3-(2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl)piperazin-2-one

  • MDL number MFCD20577119
  • Cas Number : 1373221-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135725
Availability details for
Quantity
  • Availability
  • Add to Cart