ENA408615614 Enamine

3-(1H-1,2,3-triazol-1-yl)phenol

  • MDL number MFCD26407653
  • Cas Number : 1461705-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135760
Availability details for
Quantity
  • Availability
  • Add to Cart