ENA408615619 Enamine

6-methoxy-2H-1,3-benzodioxole-5-carboxylic acid

  • MDL number MFCD16061365
  • Cas Number : 7168-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135765

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135765
Availability details for
Quantity
  • Availability
  • Add to Cart