ENA408615620 Enamine

1-[1-(2-methoxyphenyl)cyclobutyl]methanamine

  • MDL number MFCD09261725
  • Cas Number : 927993-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135766
Availability details for
Quantity
  • Availability
  • Add to Cart