ENA408615629 Enamine

4-(3-amino-4-methyl-1H-pyrazol-1-yl)benzonitrile

  • MDL number MFCD16845453
  • Cas Number : 1249427-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135777

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135777
Availability details for
Quantity
  • Availability
  • Add to Cart