ENA408615646 Enamine

4-chloro-3-hydroxypyridine-2-carboxylic acid

  • MDL number MFCD18816502
  • Cas Number : 348635-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135849
Availability details for
Quantity
  • Availability
  • Add to Cart