ENA408615647 Enamine

6-amino-1-ethyl-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD11987122
  • Cas Number : 233775-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135850

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135850
Availability details for
Quantity
  • Availability
  • Add to Cart