ENA408615654 Enamine

N-(4-bromo-3-fluorophenyl)-4-chlorobutanamide

  • MDL number MFCD21235063
  • Cas Number : 1461707-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135866

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135866
Availability details for
Quantity
  • Availability
  • Add to Cart