ENA408615677 Enamine

2-(furan-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22207169
  • Cas Number : 86423-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207-210
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121058
Availability details for
Quantity
  • Availability
  • Add to Cart