ENA408615688 Enamine

methyl 4-bromo-2-iodobenzoate

  • MDL number MFCD16877107
  • Cas Number : 1093418-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135980
Availability details for
Quantity
  • Availability
  • Add to Cart