ENA408615690 Enamine

1-(3-aminophenyl)-2-(piperidin-1-yl)ethan-1-ol

  • MDL number MFCD11593431
  • Cas Number : 55338-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135990
Availability details for
Quantity
  • Availability
  • Add to Cart