ENA408615694 Enamine

4-(3-bromophenyl)-N-(2-methoxyethyl)-1,3-thiazol-2-amine

  • MDL number MFCD17294840
  • Cas Number : 1274674-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136002
Availability details for
Quantity
  • Availability
  • Add to Cart