ENA408615703 Enamine

5'-amino-4H-[1,2'-bipyridin]-4-one

  • MDL number MFCD26407673
  • Cas Number : 1461715-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136019
Availability details for
Quantity
  • Availability
  • Add to Cart