ENA408615704 Enamine

2-(2-methoxyphenyl)-2-oxoacetic acid

  • MDL number MFCD02666071
  • Cas Number : 26767-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136020
Availability details for
Quantity
  • Availability
  • Add to Cart