ENA408615721 Enamine

2-methanesulfinylacetonitrile

  • MDL number MFCD08275182
  • Cas Number : 52109-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136065
Availability details for
Quantity
  • Availability
  • Add to Cart