ENA408615723 Enamine

2-(2,5-dimethylphenyl)ethanethioamide

  • MDL number MFCD14676744
  • Cas Number : 32669-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136067
Availability details for
Quantity
  • Availability
  • Add to Cart