ENA408615724 Enamine

4-[({[3-(pyrrolidin-1-ylmethyl)phenyl]methyl}amino)methyl]benzonitrile dihydrochloride

  • MDL number MFCD26407683
  • Cas Number : 1384715-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136068
Availability details for
Quantity
  • Availability
  • Add to Cart