ENA408615725 Enamine

N-(2-cyano-2-methylethyl)benzamide

  • MDL number MFCD16833885
  • Cas Number : 70964-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136069

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136069
Availability details for
Quantity
  • Availability
  • Add to Cart