ENA408615730 Enamine

1-(2,6-dimethylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD23160691
  • Cas Number : 1420684-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136079
Availability details for
Quantity
  • Availability
  • Add to Cart