ENA408615745 Enamine

2-(methoxymethyl)-2-methylpropane-1,3-diol

  • MDL number MFCD26085014
  • Cas Number : 21398-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136129

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136129
Availability details for
Quantity
  • Availability
  • Add to Cart