ENA408615749 Enamine

1-(phenylcarbamoyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD16669493
  • Cas Number : 145591-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136141

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136141
Availability details for
Quantity
  • Availability
  • Add to Cart