ENA408615751 Enamine

6-(piperazin-1-yl)pyridine-2-carbonitrile

  • MDL number MFCD20367055
  • Cas Number : 909413-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136143

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-136143
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart